missing translation for 'onlineSavingsMsg'
Learn More

PDIA2 Antibody, Novus Biologicals™

Product Code. 18067073 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18067073 0.025 mL 25µL
18709403 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18067073 Supplier Novus Biologicals Supplier No. NB1001921SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 6 publications

PDIA2 Polyclonal antibody specifically detects PDIA2 in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDIA2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000-1:20000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:5000-1:20000, Immunoprecipitation 1-3 ug, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:1000
Formulation Whole antisera with No Preservative
Gene Alias Pancreas-specific protein disulfide isomerase, pancreatic protein disulfide isomerase, PDA2, PDI, PDIp, PDIR, protein disulfide isomerase, pancreatic, protein disulfide isomerase-associated 2, protein disulfide-isomerase A2, Rho GDP dissociation inhibitor gamma
Gene Symbols PDIA2
Host Species Rabbit
Immunogen Purified full length PDIA2 isolated from rat liver.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, ER Markers, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 64714
Target Species Human, Mouse, Rat, Bovine, Primate
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.