Learn More
PDGFR alpha Antibody (MM0004-8A89), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60969PCP
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal antibody specifically detects PDGFR alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Protein G purified | |
| 0.1 mL |