missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE1B Antibody (10B10B12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37403
This item is not returnable.
View return policy
Description
PDE1B Monoclonal specifically detects PDE1B in Human, Mouse, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| PDE1B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| 63 kDa Cam-PDE, calcium/calmodulin-dependent 3'-5'-cyclic nucleotide phosphodiesterase 1B, Cam-PDE 1B, EC 3.1.4, EC 3.1.4.17, PDE1B1calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase, PDES1Bcalmodulin-stimulated phosphodiesterase PDE1B1, phosphodiesterase 1B, calmodulin-dependent, phosphodiesterase IB, calmodulin-dependent, Phosphodiesterase-1B, presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase familyknown as PDE1B | |
| Mouse | |
| 61.4 kDa | |
| 0.1 mL | |
| Apoptosis, Signal Transduction | |
| 5153 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 10B10B12 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PDE1B | |
| Purified recombinant fragment of human PDE1B (AA: 370-536) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Primate, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction