missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD1 Mouse anti-Canine, Human, Mouse, Rat, Clone: UMAB197, liquid, UltraMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies UM870089
This item is not returnable.
View return policy
Description
PD-1 (Programmed Cell Death Protein 1, PCDP-1, CD279) is a cell surface receptor approximately 273-290 amino acids in length that inhibits cell-mediated immune responses initiated by T lymphocytes. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of "danger signals" such as lipopolysaccharide (LPS) associated with bacteria or pathogens. Evidence for this is seen in PD1-null mice who exhibit hyper-activated immune systems and autoimmune diseases. Further, expression of this gene in mice is induced in the thymus when anti-CD3 antibodies are injected and large numbers of thymocytes undergo apoptosis. Mice deficient for this gene bred on a BALB/c background developed dilated cardiomyopathy and died from congestive heart failure.Specifications
| PDCD1 | |
| Monoclonal | |
| Unconjugated | |
| PDCD1 | |
| CD279, PD-1, PD1, SLEB2, hPD-1, hPD-l, hSLE1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 18566, 301626, 486213, 5133 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| UMAB197 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q02242, Q15116 | |
| Pdcd1 | |
| Full length human recombit protein of human PDCD1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction