Learn More
Pd(NO3)2+ for AAS Pd(NO3)2+ (conc. 2g/L), Mg(NO3)2 (conc. g/L), Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Specifications
Specifications
| Packaging | Bottle |
| Physical Form | Liquid |
| Concentration | Pd(NO3)2+ (conc. 2 g/L), Mg(NO3)2 (conc. 10 g/L) |
| Type | Matrix Modifier |
| Solvent or Matrix | 1% HNO3 matrix |
| Quantity | 50 mL |
| Chemical Name or Material | Pd(NO3)2+Mg(NO3)2 |