missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PD-L1 Antibody (opucolimab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28848-100ug
This item is not returnable.
View return policy
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| PD-L1 | |
| Monoclonal | |
| Unconjugated | |
| Q9NZQ7 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| opucolimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| B7-H1/ PD-L1/ CD274 | |
| 100 μg | |
| Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors | |
| 29126 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction