Learn More
PD-L1 Antibody (MIH5), PE/Cy5.5, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43262PECY55
This item is not returnable.
View return policy
Description
PD-L1 Monoclonal specifically detects PD-L1 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| PD-L1 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| CD274 | |
| The immunogen for this antibody was B7H1. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2a λ |
| Flow Cytometry | |
| MIH5 | |
| Flow Cytometry | |
| Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Rat | |
| Protein A or G purified | |
| Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors | |
| 29126.0 | |
| Store at 4°C in the dark. Do not freeze. |