missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-ECGF/Thymidine Phosphorylase Antibody (PGF 44C) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
214.00€
Specifications
| Antigen | PD-ECGF/Thymidine Phosphorylase |
|---|---|
| Clone | PGF 44C |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot |
Description
PD-ECGF/Thymidine Phosphorylase Monoclonal specifically detects PD-ECGF/Thymidine Phosphorylase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| PD-ECGF/Thymidine Phosphorylase | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Angiogenesis | |
| EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase | |
| TYMP | |
| IgG1 | |
| Protein A purified | |
| Cross-reacts with Human, Mouse and Rat. Not yet tested in other species. |
| PGF 44C | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 1890 | |
| Full length human protein in E. coli | |
| Primary | |
| Store at 4°C. Do not freeze. | |
| 55 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title