Learn More
Tocris Bioscience™ PD 90780
Description
PD 90780 is a a nonpeptide inhibitor of NGF binding to p75NTR (IC50 ∽ 1 μM in CHO cells); binds NGF, not p75NTR. Affinity maybe altered by the presence of TrkA receptors. Neuroprotective.
Specifications
Specifications
| Purity | >98% |
| Quantity | 50 mg |
| Product Type | PD 90780 |
| Molecular Weight (g/mol) | 362.34 |
| Inhibitors | PD 90780 |