missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PD-1 Antibody (sintilimab) - IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
213.00€ - 920.00€
Specifications
| Antigen | PD-1 |
|---|---|
| Clone | sintilimab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30046941
|
Novus Biologicals™
NBP3-28322-100ug |
100 μg |
326.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30048095
|
Novus Biologicals™
NBP3-28322-1mg |
1 mg |
920.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046519
|
Novus Biologicals™
NBP3-28322-50ug |
50 μg |
213.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| PD-1 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| PDCD1/ PD-1/ CD279 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| sintilimab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q15116 | |
| 5133 | |
| IgG4 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title