missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Antibody (J116), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
627.00€
Specifications
| Antigen | PD-1 |
|---|---|
| Clone | J116 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Biotin |
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)Specifications
| PD-1 | |
| Western Blot | |
| Biotin | |
| Mouse | |
| Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers | |
| PBS | |
| 5133 | |
| IgG1 κ | |
| Protein A or G purified |
| J116 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| The immunogen for this antibody was PD1. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title