missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Antibody (CL3624) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61625AF532
This item is not returnable.
View return policy
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| PD-1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| This antibody was developed using a recombinant protein derived from Q15116, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers | |
| 5133 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL3624 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction