missing translation for 'onlineSavingsMsg'
Learn More

PCNA Antibody (SPM350), PE, Novus Biologicals™

Product Code. 18192989 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18192989 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18192989 Supplier Novus Biologicals Supplier No. NBP234767PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PCNA Monoclonal specifically detects PCNA in Human, Mouse, Rat, Porcine, Chicken, Drosophila, Monkey, S. pombe, Yeast, Zebrafish samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCNA
Applications Flow Cytometry
Classification Monoclonal
Clone SPM350
Conjugate PE
Dilution Flow Cytometry
Gene Alias cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Gene Symbols PCNA
Host Species Mouse
Immunogen Rat PCNA/protein A fusion protein (Uniprot: P12004)
Molecular Weight of Antigen 36 kDa
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5111
Test Specificity Recognizes a non-histone protein of 36kDa, which is identified as proliferating cell nuclear antigen (PCNA). It is also known as cyclin or polymerase delta auxiliary protein. Elevated expression of PCNA/cyclin has been shown in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. This monoclonal antibody is excellent for multiple applications.
Target Species Human, Mouse, Rat, Pig, Chicken, Drosophila, Primate, Yeast, Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.