missing translation for 'onlineSavingsMsg'
Learn More

PCNA Antibody (PC10) - Azide and BSA Free, Novus Biologicals™

Product Code. 18245687 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18245687 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18245687 Supplier Novus Biologicals Supplier No. NBP280905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PCNA Monoclonal specifically detects PCNA in Human, Mouse, Rat, Porcine, Chicken, Drosophila, Fish, Primate, Rabbit, Yeast, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCNA
Applications Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone PC10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:2000, Simple Western : 1:200, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 26883631), Flow Cytometry : 1:10 - 1:1000, ELISA reported in scientific literature, Immunohistochemistry : 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:2000. Use reported in scientific literature (PMID 22068968), Immunoprecipitation : 1:100, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen : 1:10 - 1:500, Immunohistochemistry Free-Floating reported in scientific literature (PMID 27212918), Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation Tris-Glycine, 0.15M NaCl
Gene Alias cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Gene Symbols PCNA
Host Species Mouse
Immunogen Protein A-rat PCNA fusion obtained from pC2T
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5111
Test Specificity Fish reactivity reported in scientific literature (PMID: 30075024).
Target Species Human, Mouse, Rat, Pig, Chicken, Drosophila, Fish, Primate, Rabbit, Yeast, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.