missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDH9 Antibody (7G3F7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00€ - 552.00€
Specifications
| Antigen | PCDH9 |
|---|---|
| Clone | 7G3F7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18233234
|
Novus Biologicals
NBP2-61771 |
0.1 mL |
552.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677887
|
Novus Biologicals
NBP2-61771-0.025ml |
0.025 mL |
203.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCDH9 Monoclonal specifically detects PCDH9 in Human samples. It is validated for Western Blot, ELISA.Specifications
| PCDH9 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| 5101 | |
| Purified recombinant fragment of human PCDH9 (AA: 24-148) expressed in E. Coli. | |
| Primary | |
| 136 kDa |
| 7G3F7 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| protocadherin 9, protocadherin-9 | |
| PCDH9 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title