missing translation for 'onlineSavingsMsg'
Learn More

PCBD2 Antibody, HRP, Novus Biologicals™

Product Code. 18680492 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18680492 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18680492 Supplier Novus Biologicals Supplier No. NBP297231H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PCBD2 Polyclonal antibody specifically detects PCBD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCBD2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias 4-alpha-hydroxy-tetrahydropterin dehydratase 2, DCOH2DCOHMPHS2,6-pyruvoyl-tetrahydropterin synthase/dimerization cofactor of hepatocytenuclear factor 1 alpha (TCF1) 2, DcoH-like protein DCoHm, dimerization cofactor of hepatocyte nuclear factor 1 (HNF1) from muscle, dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1), Dimerization cofactor of hepatocyte nuclear factor 1 from muscle, EC 4.2.1.96, HNF-1-alpha dimerization cofactor, PHS 2, pterin-4 alpha-carbinolamine dehydratase 2, pterin-4 alpha-carbinolamine dehydratase/dimerization cofactor of hepatocytenuclear factor 1 alpha (TCF1) 2, pterin-4-alpha-carbinolamine dehydratase 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PCBD2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84105
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.