missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBEF/Visfatin/NAMPT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19468
This item is not returnable.
View return policy
Description
PBEF/Visfatin/NAMPT Polyclonal antibody specifically detects PBEF/Visfatin/NAMPT in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
| PBEF/Visfatin/NAMPT | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| EC 2.4.2.12, MGC117256, NAmPRTase, Nampt, nicotinamide phosphoribosyltransferase, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN | |
| Rabbit | |
| 56 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology, Innate Immunity | |
| 10135 | |
| Human, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P43490 | |
| NAMPT | |
| Recombinant protein encompassing a sequence within the center region of human Visfatin. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (96%), Chicken (96%), Pig (97%), Rat (96%), Xenopus laevis (92%), Bovine (96%), Xenopus tropicalis (92%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction