missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paxillin Antibody (LD4 sAB), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81063AF647
This item is not returnable.
View return policy
Description
Paxillin Monoclonal specifically detects Paxillin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Paxillin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| PXN | |
| Peptide corresponding to human paxillin-LD4 motif (residues 260-278) fused to MBP; peptide sequence: WGGSATRELDELMASLSD. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Phospho Specific, Signal Transduction | |
| 5829 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| LD4 sAB | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| FLJ16691, paxillin | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody binds specifically to the LD-4 motif of human Paxillin with a Kd of 6 nM. | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction