missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX8 Antibody (CL7834) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21161F
This item is not returnable.
View return policy
Description
PAX8 Monoclonal antibody specifically detects PAX8 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| PAX8 | |
| Monoclonal | |
| FITC | |
| paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8 | |
| This antibody was developed using a synthetic peptide derived from Q06710, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 7849 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL7834 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction