missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
218.00€
Specifications
| Antigen | Pax6 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 5 μg, Immunocytochemistry 1:100 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Pax6 Polyclonal specifically detects Pax6 in Human, Mouse, Rat, Fish, Primate, Monkey, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Pax6 | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 5 μg, Immunocytochemistry 1:100 | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers, Diabetes Research, Neuronal Stem Cell Markers, Neuronal Stem Cells, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| PBS, 1mg/ml BSA with 0.05% Sodium Azide | |
| keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
| PAX6 | |
| IgG | |
| Affinity Purified | |
| Detects a ∼47 kDa protein representing PAX6 (Schwann Cell Marker) from rat whole eye extracts and ovine retinal extracts. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P63016 | |
| 5080 | |
| Synthetic peptide corresponding to residues C R(267) E E K L R N Q R R Q A S N T P S H I(285) of mouse PAX6. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 47 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title