missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Pax5/BSAP Antibody (CL14550), Novus Biologicals™
Mouse Monoclonal Antibody
387.00€ - 582.00€
Specifications
| Antigen | Pax5/BSAP |
|---|---|
| Clone | CL14550 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:50 - 1:200 |
| Applications | Immunohistochemistry, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043781
|
Novus Biologicals™
NBP3-26865-25ul |
25 μL |
387.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043411
|
Novus Biologicals™
NBP3-26865-100ul |
100 μL |
582.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| Pax5/BSAP | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators | |
| PBS, pH 7.2, 40% glycerol | |
| 5079 | |
| IgG2a | |
| Protein A purified |
| CL14550 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
| This antibody was developed against a Synthetic Peptide of Q02548. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title