missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PATL1 Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
Specifications
Specifications
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1M Urea, pH 7.4. |
| Gene ID (Entrez) | 219988 |
| Molecular Weight (g/mol) | M.W. Theoretical: 29 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Alias | FLJ36874, hPat1b, MGC125671, OK/KNS-cl.5, Pat1b, PAT1-like protein 1, protein associated with topoisomerase II homolog 1 (yeast), protein PAT1 homolog 1, protein PAT1 homolog b |
| Gene Symbol | PATL1 |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | PATL1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction