missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvalbumin Antibody (CL15073) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43651-100ul
This item is not returnable.
View return policy
Description
Parvalbumin Monoclonal antibody specifically detects Parvalbumin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Parvalbumin | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1:5000-1:10000, Immunohistochemistry-Paraffin 1:5000-1:10000 | |
| D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV | |
| This antibody was generated using a recombinant protein sequence of P20472, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Neuroscience | |
| 5816 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL15073 | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction