missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Parvalbumin alpha Antibody (PVALB/7601), Novus Biologicals™
Mouse Monoclonal Antibody
232.00€ - 462.00€
Specifications
| Antigen | Parvalbumin alpha |
|---|---|
| Clone | PVALB/7601 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043162
|
Novus Biologicals™
NBP3-26719-100ug |
100 μg |
462.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043961
|
Novus Biologicals™
NBP3-26719-20ug |
20 μg |
232.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Parvalbumin alpha Monoclonal antibody specifically detects Parvalbumin alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Parvalbumin alpha | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| D22S749 | |
| Recombinant fragment (around aa1-110) of human Parvalbumin alpha protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| PVALB/7601 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 5816 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title