Learn More
Thermo Scientific™ Particle-Certified Glass Containers
Product Code. 15535205
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
2500 mL
4000 mL
Product Type:
Boston Round Bottle
Jug
Packer Bottle
Sample Jar
Color:
Amber
Clear
No. per Case:
4
12
24
Unit Size:
Each
Pack of 12
Pack of 24
Pack of 4
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Product Type Color No. per Case unitSize
15535205 250 mL Boston Round Bottle Clear 24 Pack of 24
15311757 60 mL Boston Round Bottle Amber 24 Pack of 24
15930371 125 mL Boston Round Bottle Amber 12 Pack of 12
16474390 125 mL Boston Round Bottle Clear 12 Pack of 12
16875781 500 mL Sample Jar Clear 12 Each
15935322 500 mL Boston Round Bottle Amber 12 Pack of 12
16484390 1000 mL Sample Jar Clear 12 Pack of 12
15926195 2500 mL Packer Bottle Amber 4 Pack of 4
15916195 4000 mL Jug Amber 4 Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15535205 Supplier Thermo Scientific™ Supplier No. 114250C/LP
Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.
Key Features:
  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
  • Individual bottles double bagged to maintain integrity until point of use
  • 4 L jugs suitable for hazardous shipping in conjunction with combination packaging
Recommended for:
  • Low particle applications
TRUSTED_SUSTAINABILITY
Specifications
Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Clear
Description Clear Glass Boston Round Bottle with Closed-top Closure, 250 mL
Diameter (English) 2.375 in.
Material Glass
No. per Case 24
Height (Metric) 136.53 mm
Product Type Boston Round Bottle
Height (English) 5.375 in.
Closure Liner PTFE/Silicone
Closure Material Polypropylene, Low-shedding
Closure Size 24-414 mm
Diameter (Metric) 60.33 mm
Closure Type Closed-top