missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP10 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00€ - 470.00€
Specifications
| Antigen | PARP10 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18676981
|
Novus Biologicals
NBP2-93286-0.02ml |
0.02 mL |
204.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668090
|
Novus Biologicals
NBP2-93286-0.1ml |
0.1 mL |
470.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PARP10 Polyclonal antibody specifically detects PARP10 in Human samples. It is validated for Western BlotSpecifications
| PARP10 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, DNA Repair, Hypoxia | |
| PBS (pH 7.3), 50% glycerol | |
| 84875 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.4.2.30, FLJ14464, PARP-10, poly (ADP-ribose) polymerase family, member 10, poly [ADP-ribose] polymerase 10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PARP10 (NP_116178.2). MVAMAEAEAGVAVEVRGLPPAVPDELLTLYFENRRRSGGGPVLSWQRLGCGGVLTFREPADAERVLAQADHELHGAQLSLRPAPPRAPARLLLQGLPPGTTPQRLEQHVQALLRASGLPVQPCCALASPRPDRALVQLPKPLSEADVRVLEEQAQNLGLEGTLVSLARVPQARAVRVVGDGASVDLLLLELYLENERRSGGGPLEDLQRLPGPLGTVASFQQWQVAERVLQQEHRLQGSELSLVPHYDILEPEELAENTSGGDHPSTQGPRATKHALLRTGGLVTALQGAGTVTMGSGEE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title