missing translation for 'onlineSavingsMsg'
Learn More

PARP (Poly ADP-Ribose Polymerase) Ab-3, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™

Epredia™ PARP (Poly ADP-Ribose Polymerase) Ab-3, Rabbit Polyclonal Antibody provides accurate, reproducible results.

159.12€ - 754.80€

Specifications

Antigen PARP (Poly ADP-Ribose Polymerase) Ab-3
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 4
Product Code Brand Quantity Purification Method Price Quantity & Availability  
Product Code Brand Quantity Purification Method Price Quantity & Availability  
12643117
View Documents
Epredia
RB-1680-P0
100 μL Purified with BSA and Azide This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
12663117
View Documents
Epredia
RB-1680-P1
500 μL Purified with BSA and Azide This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
12673117
View Documents
Epredia
RB-1680-P1ABX
500 μL Purified w/o BSA and Azide This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
12683117
View Documents
Epredia
RB-1680-PABX
1 mL Purified w/o BSA and Azide This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat and Cow. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminal region of bovine PARP

Molecular Weight: 116kDa

Positive Control: Raji cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

PARP (Poly ADP-Ribose Polymerase) Ab-3
Western Blot
Unconjugated
Rabbit
Apoptosis
A synthetic peptide from the C-terminal region of bovine PARP
Primary
1mg/mL
Polyclonal
Concentrated
RUO
Bovine, Human, Mouse
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.