Learn More
Paroxetine EP Impurity G (Paroxetine USP Related Compound E) as Hydrochloride, Mikromol™
Product Code. 18093380
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18093380 25 mg 25mg
18071380 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18093380 Supplier Mikromol Supplier No. MM1032.070025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Paroxetine EP Impurity G (Paroxetine USP Related Compound E) as Hydrochloride
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material 4-(4-Fluorophenyl)-1-methyl-1,2,3,6-tetrahydropyridine Hydrochloride
CAS 1012886-75-7
Analyte or Component Names 4-(4-Fluorophenyl)-1-methyl-1,2,3,6-tetrahydropyridine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H14 F N . Cl H
Certifications/Compliance ISO 17025
Synonym Paroxetine Hydrochloride Anhydrous Imp. G (EP), Paroxetine Imp. G (EP), 4-(4-Fluorophenyl)-1-methyl-1,2,3,6-tetrahydropyridine Hydrochloride, Paroxetine Hydrochloride Anhydrous Imp. G (EP) as Hydrochloride, Paroxetine Related Compound E as Hydrochloride, Paroxetine Hydrochloride Hemihydrate Impurity G as Hydrochloride, Paroxetine Hydrochloride Impurity G as Hydrochloride, Paroxetine Impurity G as Hydrochloride, Paroxetine Impurity G as Hydrochloride
InChI Formula InChI=1S/C12H14FN.ClH/c1-14-8-6-11(7-9-14)10-2-4-12(13)5-3-10;/h2-6H,7-9H2,1H3;1H
SMILES Cl.CN1CCC(=CC1)c2ccc(F)cc2
IUPAC Name 4-(4-fluorophenyl)-1-methyl-3,6-dihydro-2H-pyridine;hydrochloride
Molecular Weight (g/mol) 227.71
Quantity 25 mg
Formula Weight 227.0877 g/mol
Mehr anzeigen Weniger anzeigen