Learn More
Parker domnick hunter™ Clear Desiccant Cartridge for Hydrogen Gas Generators
Product Code. 12517005
Change view
Click to view available options
Type:
Desiccant Cartridge
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
12517005 Desiccant Cartridge Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12517005 Supplier Parker domnick hunter™ Supplier No. 604970050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Contains a combination of white desiccant beads and orange indicating beads that change to clear as they become saturated
  • Contains a combination of white desiccant beads and orange indicating beads that change to clear as they become saturated
  • Disposable cartridge cannot be heat regenerated
  • Vacuum sealed in aluminum foil
  • Fits all domnick hunter™ UHP Hydrogen Generator models
Specifications
For Use With (Application) Hydrogen gas generators uhp-20h, uhp-40h and uhp-60h
For Use With (Equipment) Hydrogen gas generators uhp-20h, uhp-40h and uhp-60h
Includes Combination of white desiccant beads and orange indicating beads that change to clear as they become saturated
Type Desiccant Cartridge