Learn More
Pargyline Hydrochloride, Mikromol™
Description
Pargyline Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Pargyline Hydrochloride |
| CAS | 306-07-0 |
| Analyte or Component Names | Pargyline Hydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C11 H13 N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Benzenemethanamine, N-methyl-N-2-propyn-1-yl-, hydrochloride (1:1), Benzenemethanamine, N-methyl-N-2-propynyl-, hydrochloride (9CI), Benzylamine, N-methyl-N-2-propynyl-, hydrochloride (6CI,8CI), A 19120, Eutonyl, Eutonyl-ten, MO 911, Methylbenzylpropynylamine hydrochloride, N-Benzyl-N-methyl-2-propynylamine hydrochloride, N-Methyl-N-(2-propynyl)benzylamine hydrochloride, NSC 43798, Pargyline hydrochloride |
| InChI Formula | InChI=1S/C11H13N.ClH/c1-3-9-12(2)10-11-7-5-4-6-8-11;/h1,4-8H,9-10H2,2H3;1H |
| SMILES | Cl.CN(CC#C)Cc1ccccc1 |
| IUPAC Name | N-benzyl-N-methylprop-2-yn-1-amine;hydrochloride |
| Show More |