Learn More
Pargyline Hydrochloride, LoGiCal
Product Code. 30458739
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458739 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30458739 Supplier LoGiCal Supplier No. LGCFOR1912.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pargyline Hydrochloride
CAS 306-07-0
Analyte or Component Names Pargyline Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C11 H13 N . Cl H
Certifications/Compliance ISO 9001
Synonym Benzenemethanamine, N-methyl-N-2-propyn-1-yl-, hydrochloride (1:1), Benzenemethanamine, N-methyl-N-2-propynyl-, hydrochloride (9CI), Benzylamine, N-methyl-N-2-propynyl-, hydrochloride (6CI,8CI), A 19120, Eutonyl, Eutonyl-ten, MO 911, Methylbenzylpropynylamine hydrochloride, N-Benzyl-N-methyl-2-propynylamine hydrochloride, N-Methyl-N-(2-propynyl)benzylamine hydrochloride, NSC 43798, Pargyline hydrochloride
InChI Formula InChI=1S/C11H13N.ClH/c1-3-9-12(2)10-11-7-5-4-6-8-11;/h1,4-8H,9-10H2,2H3;1H
Solution Type Neat
SMILES Cl.CN(CC#C)Cc1ccccc1
IUPAC Name N-benzyl-N-methylprop-2-yn-1-amine;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 195.69
Quantity 10 mg
Formula Weight 195.0815 g/mol