Learn More
Amcor Parafilm™ M Laboratory Film
Product Code. 11747487
Change view
Click to view available options
Length (Metric):
15.2 m
38 m
75 m
76 m
Width (Metric):
5 cm
10 cm
51 cm
Color:
Natural
Purple
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Color unitSize
11747487 15.2 m 51 cm Natural Each
10018130 76 m 10 cm Natural Each
11762644 76 m 5 cm Natural Each
11772644 38 m 10 cm Natural Each
16330422 75 m 5 cm Purple Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11747487 Supplier Amcor Supplier No. PM998
  • ONLINE EXCLUSIVE PRICE
    67.20€
    59.56€ / Pack of 1
    Save 7.64€
    11% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Efficient and cost-effective covering for test tubes, beakers, vials, petri dishes, flasks, and various other instruments and objects with irregular surfaces.
  • Protect the integrity of research applications with a tight moisture-barrier that holds a b seal and doesn't release from glassware, despite off-gassing
  • Reduce the risk of tampered data with a covering that is odorless, colorless and semi-transparent
  • Prevents moisture and volume loss due to material composition and tight seal
  • Flexible and self-sealing material allows it to cling around irregular shapes and surfaces
  • Stretches more than 200% of original length
  • Resistant up to 48 hours against many polar substances such as saline solutions, inorganic acids and alkaline solutions
Permeability
  • Oxygen: (ASTM 1927-98) 150 cc/m2 d at 23°C and 50% RH
  • Carbon Dioxide: (Modulated IR Method) 1200 cc/m2 d at 23°C and 0% RH
  • Water Vapor: (ASTM F1249-01)
    • Flat: 1 g/m2 d at 38°C and 90% RH
    • Creased: 1 g/m2 d at 38°C and 90% RH
Effects of Common Reagents
  • Potassium Permanganate
    • 5%: No apparent effect except permanent dark brown coloration in 18 hours
    • 0.1%: Same as 5% except slightly less color
  • Iodine solution (0.1N)
    • No effect except staining brown in 18 hours
  • Ethyl Alcohol (95%)
    • No apparent effect except some face whitening in 24 hours
  • No apparent effect in 24 hours:
    • Hydrochloric Acid conc. (12N) dil. (5N)
    • Sulphuric Acid conc. (36N) dil. (5N)
    • Nitric Acid conc. (16N) dil. (5N)
    • Sodium Hydroxide conc. (22%)
    • Ammonium Hydroxide conc. (28% NH3)
    • Salt (NaCl) solution (20%)
    • Isopropyl Alcohol (99%)
  • Not recommended for use with chlorinated, non-polar aliphatic and aromatic solvents
  • Film becomes soft and sticky at about 130° to 150°F (54° to 66°C)
Order Info
13-374-10, 13-374-12, and 13-374-16 rolls are packaged in cardboard dispensing cartons.
Specifications
Applications Efficient and cost-effective covering, over wrap to further secure stoppers, lids, and caps, and shelf or tray liner to prevent slipping of bottles or instruments.
Length (English) 50 ft.
Color Natural
Description Amcor PM998
Product Type Film
Width (English) 20 in.
Width (Metric) 51 cm
Length (Metric) 15.2 m
Material Thermoplastic
Thickness 5 mil
For Use With (Equipment) Parafilm Wrap