missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pappalysin-1/PAPP-A Mouse anti-Human, Clone: 9, Novus Biologicals™
Mouse Monoclonal Antibody
506.00€
Specifications
| Antigen | Pappalysin-1/PAPP-A |
|---|---|
| Clone | 9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:8000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen |
| Applications | Western Blot |
Description
Pappalysin-1/PAPP-A Monoclonal specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Pappalysin-1/PAPP-A | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| 5069 | |
| IgG1 κ | |
| Protein A or G purified |
| 9 | |
| Western Blot 1:100-1:2000, ELISA 1:8000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1 | |
| Purified human PAPP-A | |
| Primary | |
| Store at 4°C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title