Learn More
Epredia™ Papanicolaou OG-6 and Modified OG Stains
Product Code. p-4530219 Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
Type:
OG-6 Papanicolaou Stain
Unit Size:
3.79 L
Product Code. 10444340
Brand: Epredia™ 75211
Stain gynecological specimens with Epredia™ Papanicolaou Stain OGs.
  • OG-6 stains the keratinized areas of the cell
  • Modified OG is a Gill modification that minimizes precipitation and reduces staining time
  • Use OG-6 with EA-36 or EA-50 when staining gynecological specimens
  • Use modified OG with EA-65 for staining non-gynecological specimens
TRUSTED_SUSTAINABILITY
Specifications
Type OG-6 Papanicolaou Stain
Quantity 1 gal.