missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Papain Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-20595-0.1mg
This item is not returnable.
View return policy
Description
Papain Polyclonal antibody specifically detects Papain in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Papain | |
| Polyclonal | |
| Biotin | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
| Allergen Car p 1, Papaya proteinase I, PPI | |
| Papain Antibody was produced by repeated immunizations with pork Papain protein isolated from papaya. (Uniprot: P00784) | |
| 0.1 mg | |
| Primary | |
| Plant | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:500-1:2000, ELISA 1:2000-1:20000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| P00784 | |
| Goat | |
| Multi-step | |
| RUO | |
| Reconstitute with 100 μg deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction