Learn More
STEMCELL Technologies Papain
Product Code. 17129281
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17129281 100 mg 100mg
17119281 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17129281 Supplier STEMCELL Technologies Supplier No. 07466
For digestion of the extracellular matrix of cartilage
  • Papain is a cysteine protease consisting of a single polypeptide containing three disulfide bridges
  • Native crystalline papain is unreactive until acted upon by mild reducing agents, e.g. , cysteine, sulfide, or sulfite, and therefore likely exists as a zymogen
  • Papain has a wide specificity with a preference towards arginine, lysine, and phenylalanine
  • This enzyme degrades protein substrates such as the intercellular matrices of cartilage more extensively than pancreatic proteases and is typically less damaging and more effective than other proteases for tissue dissociation applications (Huettner & Baugham; Lam) and has also been used for the digestion of neural tissue (Fasano et al.)
Specifications
Description Dopaminergic Neurons; Mesenchymal Cells, PSC-Derived; Neurons; Pancreatic Cells
Quantity 100 mg
Synonym Papainase; Papaya peptidase I