missing translation for 'onlineSavingsMsg'
Learn More

Pancreatic Amylase Alpha Antibody (OTI1E3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-73239F

Product Code. 18164976

  • 751.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Pancreatic Amylase Alpha Monoclonal specifically detects Pancreatic Amylase Alpha in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pancreatic Amylase Alpha
Monoclonal
FITC
PBS with 0.05% Sodium Azide
AMY2A
Recombinant protein fragment corresponding to amino acids 135-408 of human Pancreatic Amylase Alpha (NP_000690) produced in E.coli.
0.1 mL
279.0
Store at 4°C in the dark.
Immunohistochemistry
OTI1E3
Immunohistochemistry
AMY2, AMY2A amylase, alpha 2A (pancreatic), AMY2B, PA, pancreatic alpha-amylase
Mouse
Affinity Purified
Primary
Human
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.