missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pan-Synuclein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
302.00€ - 456.00€
Specifications
| Antigen | pan-Synuclein |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18160118
|
Novus Biologicals
NBP2-47338 |
0.1 mL |
456.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649265
|
Novus Biologicals
NBP2-47338-25ul |
25 μL |
302.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
pan-Synuclein Polyclonal specifically detects pan-Synuclein in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| pan-Synuclein | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Cell Biology, Cytoskeleton Markers, Neurodegeneration, Neuroscience, Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 6622 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EGVVHGVATVAEKTKEQVTNVGGAVVTGVTAVAQKTVEGAGSIAAATGFVKKDQLGKNEEGAPQEGILEDMPVDPDNEAYEMPSEEGYQDYEPE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK4, synuclein, alpha (non A4 component of amyloid precursor) | |
| SNCA | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title