missing translation for 'onlineSavingsMsg'
Learn More

pan-keratin Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen pan-keratin
Concentration 0.3 mg/mL
Applications Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16824885
View Documents
Proteintech
26411-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16814885
View Documents
Proteintech
26411-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is a pan-keratin antibody.

The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.
Specifications

Specifications

pan-keratin
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
P02533, P02538, P04264, P05787, P08727, P08729, P08779, P12035, P13645, P13647, P19012, P19013, P35908
3848, 3849, 3850, 3851, 3852, 3853, 3855, 3856, 3858, 3861, 3866, 3868, 3880
KRT5 Fusion Protein Ag24184
Primary
-20°C
KRT10
0.3 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
58 kDa cytokeratin, CK 5, CK5, Cytokeratin 5, K5, keratin 5, KRT5, KRT5A, Type II keratin Kb5
KRT1, KRT10, KRT14, KRT15, KRT16, KRT19, KRT2, KRT3, KRT4, KRT5, KRT6A, KRT7, KRT8
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.