missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pan Actin Antibody (AC-40), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
221.00€
Specifications
| Antigen | pan Actin |
|---|---|
| Clone | AC-40 |
| Dilution | Western Blot 1:500, Simple Western, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
pan Actin Monoclonal specifically detects pan Actin in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Guinea Pig, Goat, Primate, Rabbit, Reptile, Sheep samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| pan Actin | |
| Western Blot 1:500, Simple Western, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3 | |
| ACTA1 | |
| IgG2a | |
| Unpurified | |
| The does not react with the epitope on the N-terminal end of actin. |
| AC-40 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers | |
| Ascites with 15mM Sodium Azide | |
| 58 | |
| Synthetic peptide: SGPSIVHALCP (human) attached to Multiple Antigen Peptide (MAP) backbone. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 42 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title