missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDH1 Mouse anti-Human, Clone: CPTC-MDH1-1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07289-100ug
This item is not returnable.
View return policy
Description
MDH1 Monoclonal specifically detects MDH1 in Human samples. It is validated for Western Blot.
Specifications
| MDH1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL | |
| cytoplasmic, Cytosolic malate dehydrogenase, DEE88, Diiodophenylpyruvate reductase, EC 1.1.1.37, EC 1.1.1.96, HEL-S-32, malate dehydrogenase 1, NAD (soluble), Malate dehydrogenase, cytoplasmic, MDH1, MDHA, MDH-s, MGC:1375, MOR2, soluble malate dehydrogenase | |
| Recombinant human full-length MDH1 protein | |
| 100 μg | |
| Cancer, Immunology, Stem Cell Markers, Virology Bacteria and Parasites | |
| 4190 | |
| Store at 4°C. | |
| IgG2a κ |
| Western Blot | |
| CPTC-MDH1-1 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction