missing translation for 'onlineSavingsMsg'
Learn More

PALB2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

178.00€

Specifications

Antigen PALB2
Concentration 1 mg/mL
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18522431
View Documents
Novus Biologicals
NBP1-76317-0.025mg
0.025 mg
178.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PALB2 Polyclonal specifically detects PALB2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

PALB2
Western Blot 1-2 μg/mL, ELISA 1:100-1:2000
Polyclonal
Purified
RUO
Human
Q86YC2
79728
Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human PALB2. The immunogen is located within amino acids 940 - 990 of PALB2. Amino Acid Squence: SDDESEKQVLLKSGN
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
140 kDa
1 mg/mL
Western Blot
Unconjugated
Rabbit
Breast Cancer, Cancer
PBS with 0.02% Sodium Azide
DKFZp667I166, DKFZp686E1054, FANCNFLJ21816, partner and localizer of BRCA2, PNCA3
PALB2
IgG
Peptide affinity purified
At least four isoforms of PALB2 are known to exist; this antibody will detect all four isoforms.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.