missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK2 Antibody (3B5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47542SS
This item is not returnable.
View return policy
Description
PAK2 Monoclonal specifically detects PAK2 in Human, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PAK2 | |
| Monoclonal | |
| Unconjugated | |
| EC 2.7.11, gamma-PAK, p21 (CDKN1A)-activated kinase 2, p21 protein (Cdc42/Rac)-activated kinase 2, p21-activated kinase 2, p58, PAK-2, PAK65EC 2.7.11.1, PAKgamma, S6/H4 kinase, serine/threonine-protein kinase PAK 2 | |
| Mouse | |
| 61 kDa | |
| 0.025 mL | |
| Protein Kinase | |
| 5062 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 3B5 | |
| Western Blot 1: 500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| PAK2 | |
| Purified recombinant fragment of PAK2 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction