missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PAFAH1B3 (Human) Recombinant Protein
Human PAFAH1B3 full-length ORF ( AAH03016, 1 a.a. - 231 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005050-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSGEENPASKPTPVQDVQGDGRWMSLHHRFVADSKDKEPEVVFIGDSLVQLMHQCEIWRELFSPLHALNFGIGGDGTQHVLWRLENGELEHIRPKIVVVWVGTNNHGHTAEQVTGGIKAIVQLVNERQPQARVVVLGLLPRGQHPNPLREKNRQVNELVRAALAGHPRAHFLDADPGFVHSDGTISHHDMYDYLHLSRLGYTPVCRALHSLLLRLLAQDQGQGAPLLEPAP
Specifications
AAH03016 | |
platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit 29kDa | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5050 | |
Wheat germ expression system | |
10 μg | |
PAFAH1B3 | |
GST |