missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PAFAH1B3 (Human) Recombinant Protein

Human PAFAH1B3 full-length ORF ( AAH03016, 1 a.a. - 231 a.a.) recombinant protein with GST-tag at N-terminal.

335.00€ - 508.00€

Specifications

Accession Number AAH03016
Gene ID (Entrez) 5050
Name platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit 29kDa
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16148671
View Documents Promotion Details
Abnova™
H00005050-P01.10UG
10 μg
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16158671
View Documents Promotion Details
Abnova™
H00005050-P01.25UG
25 μg
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

  • Sequence: MSGEENPASKPTPVQDVQGDGRWMSLHHRFVADSKDKEPEVVFIGDSLVQLMHQCEIWRELFSPLHALNFGIGGDGTQHVLWRLENGELEHIRPKIVVVWVGTNNHGHTAEQVTGGIKAIVQLVNERQPQARVVVLGLLPRGQHPNPLREKNRQVNELVRAALAGHPRAHFLDADPGFVHSDGTISHHDMYDYLHLSRLGYTPVCRALHSLLLRLLAQDQGQGAPLLEPAP
Specifications

Specifications

AAH03016
platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit 29kDa
125% SDS-PAGE Stained with Coomassie Blue
PAFAH1B3
GST
5050
Wheat germ expression system
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Product Certifications