missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAEP/Glycodelin Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18187-1-AP-20UL
This item is not returnable.
View return policy
Description
The PAEP/Glycodelin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PAEP/Glycodelin. This antibody recognizes human antigen. The PAEP/Glycodelin antibody has been validated for the following applications: IHC, ELISA analysis.Specifications
| PAEP/Glycodelin | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5047 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Immunohistochemistry, ELISA | |
| 300 μg/mL | |
| GD; GdA; GdF; GdS; MGC138509; MGC142288; PAEG; PEP; PP14 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 24-28 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction