missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p70 S6 Kinase/S6K Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 1 publication
213.00€
Specifications
| Antigen | p70 S6 Kinase/S6K |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Peptide ELISA 1:100 - 1:1000, ICC/IF (Negative) |
| Applications | Western Blot, ELISA, Immunocytochemistry |
| Classification | Polyclonal |
Description
p70 S6 Kinase/S6K Polyclonal specifically detects p70 S6 Kinase/S6K in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, Peptide ELISA, ICC/IF (Negative).Specifications
| p70 S6 Kinase/S6K | |
| Western Blot 1:100 - 1:2000, Peptide ELISA 1:100 - 1:1000, ICC/IF (Negative) | |
| Polyclonal | |
| Goat | |
| Autophagy, Phospho Specific, Protein Kinase, Signal Transduction | |
| Whole sera with 50% Glycerol with 0.025% Sodium Azide | |
| 6198 | |
| Synthetic peptide made to a C-terminal sequence of the human p70 S6 Kinase protein (between amino acids 350-472). [UniProt P23443-2]. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Bovine, Rabbit | |
| EC 2.7.11, EC 2.7.11.1, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6 kinase, alpha 1,70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase, alpha 2, p70 S6KA, p70 S6K-alpha, p70(S6K)-alpha, p70-alpha, p70-S6K, P70S6K1, PS6K, ribosomal protein S6 kinase beta-1, Ribosomal protein S6 kinase I, ribosomal protein S6 kinase, 70kD, polypeptide 1, ribosomal protein S6 kinase, 70kDa, polypeptide 1, S6K, S6K1p70-S6K 1, S6K-beta-1, serine/threonine kinase 14 alpha, Serine/threonine-protein kinase 14A, STK14A | |
| RPS6KB1 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey and chimpanzee, 94% homologous in mouse and rat, 87% homologous in dog and 81% homologous in horse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title