missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p70 S6 Kinase beta/S6K2, p Ser371 Antibody (S01-4A5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19549-100ul
This item is not returnable.
View return policy
Description
p70 S6 Kinase beta/S6K2 Monoclonal antibody specifically detects p70 S6 Kinase beta/S6K2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| p70 S6 Kinase beta/S6K2 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S01-4A5 | |
| Unconjugated | |
| EC 2.7.11, EC 2.7.11.1, KLS, p70 ribosomal S6 kinase beta, p70 S6 kinase beta, p70 S6KB, p70 S6K-beta, p70(S6K)-beta, p70-beta, P70-beta-1, P70-beta-2, p70-S6K 2, P70S6K2, p70S6Kb, ribosomal protein S6 kinase beta-2, ribosomal protein S6 kinase, 70kD, polypeptide 2,70 kDa ribosomal protein S6 kinase 2, ribosomal protein S6 kinase, 70kDa, polypeptide 2, S6K2, S6K-beta, S6K-beta2, S6K-beta-2, serine/threonine-protein kinase 14 beta, Serine/threonine-protein kinase 14B, SRK, STK14BS6 kinase-related kinase | |
| A synthetic phosphopeptide corresponding to residues surrounding Ser371 of human p70 S6 Kinase beta/S6K2 (Uniprot # Q9UBS0) | |
| 100 μg | |
| Autophagy, Cancer, Cell Cycle and Replication, Phospho Specific | |
| 6199 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction