missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p63/TP73L Antibody (CL3748), Novus Biologicals™
Mouse Monoclonal Antibody
338.00€ - 662.00€
Specifications
| Antigen | p63/TP73L |
|---|---|
| Clone | CL3748 |
| Dilution | Western Blot 1 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661247
|
Novus Biologicals
NBP3-05501-100ul |
100 μg |
662.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654688
|
Novus Biologicals
NBP3-05501-25ul |
25 μg |
338.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p63/TP73L Monoclonal antibody specifically detects p63/TP73L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| p63/TP73L | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| amplified in squamous cell carcinoma, Chronic μgcerative stomatitis protein, CUSP, EEC3B(p51B), Keratinocyte transcription factor KET, KETRHS, LMS, OFC8SHFM4TP73LNBP, p40, p51tumor protein p53-like, p53CP, P63, p63AIS, p73H, P73L, p73LB(p51A), TP53CP, TP53L, Transformation-related protein 63, tumor protein 63, tumor protein p53-competing protein, tumor protein p63, tumor protein p63 deltaN isoform delta, Tumor protein p73-like | |
| This antibody was developed against a recombinant protein corresponding to amino acids: CACPGRDRKADEDSIRKQQVSDSTKNGDAFRQNTHGIQMTSIKKRRSPDDELLYLPVRGRETYEMLLKIKESLELMQYLPQHTIETYRQQQQQQHQ | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL3748 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia, Stem Cell Markers, Tumor Suppressors | |
| PBS, pH 7.2, containing 40% glycerol | |
| 8626 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title