missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 Tumor Suppressor Protein Mouse, Clone: PCRP-TP53-2A10, enQuireBio™
Mouse Monoclonal Antibody
199.00€ - 478.00€
Specifications
| Antigen | p53 Tumor Suppressor Protein |
|---|---|
| Clone | PCRP-TP53-2A10 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15901008
|
EnquireBio
7157-MSM14-P0 |
20 μg | |||||||
|
15911008
|
EnquireBio
7157-MSM14-P1 |
100 μg |
573.00€ 478.00€ / 100µg Save 95.00€ 16% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| p53 Tumor Suppressor Protein | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human TP53 protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 53kDa |
| PCRP-TP53-2A10 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 7157 | |
| IgG2b | |
| Protein A/G purified from Bioreactor concentrate. | |
| Nuclear | |
| HeLa cells. Colorectal or Ovarian Carcinoma. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title